Thermo Scientific PH480X1 Hearth Plate, Ceramic
Representative image only.
Thermo Scientific PH480X1 Hearth Plate, Ceramic
Thermo Scientific | Mfr: PH480X1| Item: 0205360
₹33,500.00INR / Each
₹33,500.00INR / Each
Buy It Together
Current Item
Thermo Scientific PH480X1 Hearth Plate, Ceramic
Thermo Scientific PH480X1 Hearth Plate, Ceramic
0205360
₹33,500.00INR / Each
+ + + =
Total Price 2104500.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Ceramic hearth plates, shelves and other parts and accessories for your muffle furnace.
Specifications
  • Capacity (cu in)350
  • Max Temperature (° C)1200
  • DescriptionHearth Plate, Ceramic