Thermo Scientific Nalgene 4250-0100 polypropylene analytical funnel, 254 mL

Representative image only.

Thermo Scientific Nalgene 4250-0100 polypropylene analytical funnel, 254 mL

Thermo Scientific | Mfr: 4250-0100| Item: 0612060

No air lock with outer ribs

₹12,800.00INR / Pkg of 4

₹12,800.00INR / Pkg of 4

Buy It Together

Current Item

Thermo Scientific Nalgene 4250-0100 polypropylene analytical funnel, 254 mL
Thermo Scientific Nalgene 4250-0100 polypropylene analytical funnel, 254 mL
0612060

₹12,800.00INR / Pkg of 4
+
Disposable PP Vented Utility Funnel, 40 mL; 100/Bag
0612410

₹6,600.00INR / Bag of 100
+
Cole-Parmer Utility Funnels, PP, 40 mL; 12/PK
6310030

₹4,400.00INR / Pkg of 12
=
Total Price 23800.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Get faster filtration with 58° angled internal ribs
  • Accepts standard filter papers
  • Autoclavable
  • 1 year warranty

More About this Item

These funnels can use standard filter paper. Long stems and external ribbing eliminates air lock. The funnels are shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. The body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration.

1 Year Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Capacity (mL)254
  • Capacity (oz)8.625
  • Stem OD (mm)9
  • Top OD (in)4 1/4
  • Top OD (mm)108
  • Filter Paper Diameter (mm)185
  • Descriptionpolypropylene analytical funnel, 254 mL
  • Warranty1 year
Documents & Certificates
Reviews
Q&A