Thermo Scientific Nalgene 300-4000 Polysulfone Filter Holder and Receiver, 250/250 mL
Representative image only.
Thermo Scientific Nalgene 300-4000 Polysulfone Filter Holder and Receiver, 250/250 mL
Thermo Scientific | Mfr: 300-4000| Item: 0673050
Versatile system with interchangeable components
₹50,400.00INR / Each
₹50,400.00INR / Each
Buy It Together
Current Item
Thermo Scientific Nalgene 300-4000 Polysulfone Filter Holder and Receiver, 250/250 mL
Thermo Scientific Nalgene 300-4000 Polysulfone Filter Holder and Receiver, 250/250 mL
0673050
₹50,400.00INR / Each
+ =
Total Price 133100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Autoclavable, break-resistant, and nontoxic
  • Exhibits low protein binding
  • Contains low levels of trace metals and organic leachables
  • 1 year warranty
More About this Item
Polysulfone (PSF) filter holders have an upper chamber with either a receiver or funnel. Choose from 250- or 500-mL upper chambers and either 250-, 500-, or 1000-mL receivers (all are graduated in 50-mL increments). Holders accept 47- and 50-mm diameter membranes. Supplied with two types of membrane supports. Use the sterilization membrane support for fast flow through; use the analytical support to keep the membrane perfectly flat for the analysis of trapped particles. Receivers with covers are available separately to increase your storage capacity. Use holders for vacuum or pressure filtration up to 10 psig. All ports accept 1/4" to 5/16" ID pressure tubing. Funnel models have vacuum gaskets to fit filtering flasks.
1 Year Warranty
Specifications
  • Body MaterialPolysulfone
  • Receiver Capacity (mL)250
  • Funnel Size (mL)250
  • Upper Chamber Capacity (mL)250
  • DescriptionPS Filter Holder and Receiver, 250/250 mL