Thermo Scientific Nalgene Reusable Polysulfone Bottletop Filter Holders, 45-mm Neck
Removable membrane support plate is designed to provide maximum flow rate and throughput
  • Autoclavable
  • Noncytotoxic
These Nalgene® polysulfone (PSF) bottletop filters screw directly onto glass media bottles with 45-mm neck size. Deep threads hold the filter onto the bottles. Use with 47-mm diameter membranes. Nominal filter area is 13.3 cm2 with sterilization support plate.
PSF bottletop filters feature a vacuum port, molded-in graduations, and silicone O-ring. For a sterile venting plug, put cotton in the polypropylene tubing adapter. Filters accept 1/4" to 5/16" (6- to 8-mm) ID vacuum tubing.
82,700.00 - ₹109,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Funnel Size (mL)
Availability Pricing
Thermo Scientific Nalgene DS0320-2545 PS Bottletop Filter Holder, 47 mm, 250 mL 3040802 Mfr # DS0320-2545
COMPARE
47 250
₹82,700.00INR / Each
Thermo Scientific Nalgene DS0320-5045 PS Bottletop Filter Holder, 45 mm, 500 mL; 1/Pk 3040812 Mfr # DS0320-5045
COMPARE
45 500
₹109,000.00INR / Each