Thermo Scientific D0809 Deionization Cartridge, high purity, 875 grain
Representative image only.
Thermo Scientific D0809 Deionization Cartridge, high purity, 875 grain
Thermo Scientific | Mfr: D0809| Item: 0150526
Keep systems operating precisely
₹54,700.00INR / Each
₹54,700.00INR / Each
Product Description
Key Features
  • Ideal for high purity with lower capacity
  • Use for most analytical lab testing
  • 1 year warranty
More About this Item
These high capacity cartridge remove ionized impurities to produce high resistivity water with a neutral pH.
Model 01505-24 is ideal for high capacity, lower purity applications and compatible with the B-Pure, E-Pure, all Nanopure, and Bantam Barnstead systems.
Model 01506-26 and 01505-29 are suitable for high purity, lower capacity usage. The model 01506-26 cartridge is compatible with the B-Pure, Nanopure II, old Nanopure, and Bantam systems, while model 01505-29 is compatible with E-Pure and quad-style Nanopure.
1 Year Warranty
Specifications
  • Capacity (grains)875 as Calcium Carbonate
  • Compatible withNANOpure™ II, old NANOpure™, B-pure™
  • FunctionHigh purity, lower capacity; use for most analytical lab testing
  • DescriptionDeionization Cartridge, high purity, 875 grain