Thermo Scientific D0760 Deionization Cartridge, anion removal
Representative image only.
Thermo Scientific D0760 Deionization Cartridge, anion removal
Thermo Scientific | Mfr: D0760| Item: 0150516
₹97,400.00INR / Each
₹97,400.00INR / Each
Buy It Together
Current Item
Thermo Scientific D0760 Deionization Cartridge, anion removal
Thermo Scientific D0760 Deionization Cartridge, anion removal
0150516
₹97,400.00INR / Each
+
Thermo Scientific Barnstead D0800 Bantam Deionizer, 120V
9926101
₹436,000.00INR / Each
=
Total Price 533400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Selectable cartridges to best meet your application
  • 2 year warranty
More About this Item
Disposable cartridges are easily replaced by turning a thumbscrew.
Two year warranty
Specifications
  • Capacity (grains)1680 as Sodium Chloride
  • FunctionRemoval of weakly ionized impurities, raises the pH of solutions, recovers precious metal complex
  • DescriptionDeionization Cartridge, anion removal