Thermo Scientific COD165 Reactor for COD test, 115/230VAC
Representative image only.
Thermo Scientific COD165 Reactor for COD test, 115/230VAC
Thermo Scientific | Mfr: COD165| Item: 9958135
Ideal for environmental and wastewater monitoring
₹456,000.00INR / Each
₹456,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific COD165 Reactor for COD test, 115/230VAC
Thermo Scientific COD165 Reactor for COD test, 115/230VAC
9958135
₹456,000.00INR / Each
+ + + =
Total Price 572000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Simultaneous digestion of 25 samples saves time
  • For a complete system order COD reactor (99581-35), COD test kits (99581-40,-42, or -44), and COD colorimeter (99581-20)
  • 1 year warranty
More About this Item
Works with Thermo Scientific Orion COD kits or any 16 mm COD vial. The simple two-button interface, combined with the auto-shutoff feature, easily sets the reactor to digest up to 25 vials at once. Preset reaction times of 30, 60, or 120 minutes eliminate the need for an external timer. Use continuous mode for other digestion times. Set the temperature to 100°C, 120°C, 150°C, 160°C, or 165°C to start the reactor.
1 Year WarrantyISO9001:2000 Certified Supplier
Specifications
  • Meter TypeAccessory; Reactors
  • DescriptionReactor for COD test, 115/230VAC