Environmental Express® HotBlock COD Reactor
Digest two analytical batches at once!
  • Process up to fifty-six standard, 16-mm diameter, round-bottom COD vials
  • See actual and set-point temperatures simultaneously
  • Pre-programmed to run at 150°C for two hours, and flashes when the cycle is complete
  • Maintain a setpoint of ±0.1°C and sample consistency of ±1.5°C
Reliable and uniform HotBlock technology now in a COD reactor. Process up to 56 samples, two analytical batches, in a single run! The digital display shows the actual temperature (in red) and set temperature (in green) simultaneously.
Units come pre-programmed to run at 150°C for two hours for standard COD digestions, but may be adjusted in 0.1°C increments from ambient to 150°C for other reactions . The heating mat maintains a setpoint within ±0.1°C and sample consistency of ±1.5°C. The Safe-Sample™ temperature protection system alerts you in the event of runaway temperatures. The unit will sound an alarm and shut off heating to protect samples.
1,294,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test Description
Availability Pricing
COMPARE
Ambient to 150°C, with 0.1° resolution and ±1.5°C accuracy Chemical oxygen demand (COD) COD Reactor, 56-Place; 115 VAC
Call For Price
COMPARE
Ambient to 150°C, with 0.1° resolution and ±1.5°C accuracy Chemical oxygen demand (COD) COD Reactor, 56-Place; 240 VAC
₹1,294,000.00INR / Each