Thermo Scientific Barnstead D8950 Ion Exchange Cartridge; 2/3-Size, Two-Bed
Representative image only.
Thermo Scientific Barnstead D8950 Ion Exchange Cartridge; 2/3-Size, Two-Bed
Thermo Scientific | Mfr: D8950| Item: 9926003
Customize your system with application-specific water purification cartridges
₹41,000.00INR / Each
₹41,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D8950 Ion Exchange Cartridge; 2/3-Size, Two-Bed
Thermo Scientific Barnstead D8950 Ion Exchange Cartridge; 2/3-Size, Two-Bed
9926003
₹41,000.00INR / Each
+ +
Thermo Scientific Barnstead D50229 cartridge, 10"L x 2" dia
9926152
₹62,400.00INR / Each
+ =
Total Price 166800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 2 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
Color-changing resin indicates when cartridge is exhausted. Cannot be used with alcohol-containing fluids.
Two year warranty
Specifications
  • MediaResin
  • Capacity (grains)1100 as Sodium Chloride
  • Connection3/8" hose barb at each end
  • Functionhigh capacity, ion exchange, strong cation exchange
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionIon Exchange Cartridge; 2/3-Size, Two-Bed