Thermo Scientific Barnstead D8911 Disposable Ion Exchange Cartridge, ultrapure
Representative image only.
Thermo Scientific Barnstead D8911 Disposable Ion Exchange Cartridge, ultrapure
Thermo Scientific | Mfr: D8911| Item: 9926010
Customize your system with application-specific water purification cartridges
₹50,900.00INR / Each
₹50,900.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D8911 Disposable Ion Exchange Cartridge, ultrapure
Thermo Scientific Barnstead D8911 Disposable Ion Exchange Cartridge, ultrapure
9926010
₹50,900.00INR / Each
+ +
Thermo Scientific Barnstead D50229 cartridge, 10"L x 2" dia
9926152
₹62,400.00INR / Each
+ =
Total Price 176700.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 2 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
Two year warranty
Specifications
  • MediaResin
  • Capacity (grains)915 as Sodium Chloride
  • Connection3/8" hose barb at each end
  • FunctionUltrapure ion exchange
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionDisposable Ion Exchange Cartridge, ultrapure