Thermo Scientific Barnstead D8904 Ion Exchange Cartridge, active charcoal
Representative image only.
Thermo Scientific Barnstead D8904 Ion Exchange Cartridge, active charcoal
Thermo Scientific | Mfr: D8904| Item: 9926008
Customize your system with application-specific water purification cartridges
₹39,000.00INR / Each
₹39,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D8904 Ion Exchange Cartridge, active charcoal
Thermo Scientific Barnstead D8904 Ion Exchange Cartridge, active charcoal
9926008
₹39,000.00INR / Each
+
Thermo Scientific Barnstead D50229 cartridge, 10"L x 2" dia
9926152
₹62,400.00INR / Each
+ + =
Total Price 164800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 1 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
1 Year Warranty
Specifications
  • MediaActivated charcoal
  • Connection3/8" hose barb at each end
  • FunctionOrganics and chlorine removal
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionIon Exchange Cartridge, active charcoal