Thermo Scientific Barnstead D5027 Deionization Cartridge, high purity, 760 grain
Representative image only.
Thermo Scientific Barnstead D5027 Deionization Cartridge, high purity, 760 grain
Thermo Scientific | Mfr: D5027| Item: 0150529
Keep systems operating precisely
₹53,400.00INR / Each
₹53,400.00INR / Each
Product Description
Key Features
  • Ideal for high purity with lower capacity
  • Use for most analytical lab testing
More About this Item
These high capacity cartridge remove ionized impurities to produce high resistivity water with a neutral pH.
Model 01505-24 is ideal for high capacity, lower purity applications and compatible with the B-Pure, E-Pure, all Nanopure, and Bantam Barnstead systems.
Model 01506-26 and 01505-29 are suitable for high purity, lower capacity usage. The model 01506-26 cartridge is compatible with the B-Pure, Nanopure II, old Nanopure, and Bantam systems, while model 01505-29 is compatible with E-Pure and quad-style Nanopure.
Specifications
  • Capacity (grains)760 as Sodium Chloride
  • Compatible withQuad-style NANOpure™, E-pure™
  • FunctionHigh purity, lower capacity; use for most analytical lab testing
  • DescriptionDeionization Cartridge, high purity, 760 grain