Thermo Scientific Barnstead D0815 Deionization Cartridge, cation removal
Representative image only.
Thermo Scientific Barnstead D0815 Deionization Cartridge, cation removal
Thermo Scientific | Mfr: D0815| Item: 0150518
₹41,300.00INR / Each
₹41,300.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D0815 Deionization Cartridge, cation removal
Thermo Scientific Barnstead D0815 Deionization Cartridge, cation removal
0150518
₹41,300.00INR / Each
+
Thermo Scientific Barnstead D0800 Bantam Deionizer, 120V
9926101
₹436,000.00INR / Each
=
Total Price 477300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Selectable cartridges to best meet your application
  • 2 year warranty
More About this Item
Disposable cartridges are easily replaced by turning a thumbscrew.
Two year warranty
Specifications
  • Capacity (grains)3170 as Sodium Chloride
  • FunctionConverts ionized salts to the acid from resulting in a product water that is low in pH, ideal for precious metal or isotope recovery
  • DescriptionDeionization Cartridge, cation removal