Thermo Scientific Barnstead D0803 Deionization Cartridges, high capacity
Representative image only.
Thermo Scientific Barnstead D0803 Deionization Cartridges, high capacity
Thermo Scientific | Mfr: D0803| Item: 0150524
Keep systems operating precisely
₹50,500.00INR / Each
₹50,500.00INR / Each
Product Description
Key Features
  • Ideal for high purity with lower capacity
  • Use for most analytical lab testing
  • 1 year warranty
More About this Item
These high capacity cartridge remove ionized impurities to produce high resistivity water with a neutral pH.
Model 01505-24 is ideal for high capacity, lower purity applications and compatible with the B-Pure, E-Pure, all Nanopure, and Bantam Barnstead systems.
Model 01506-26 and 01505-29 are suitable for high purity, lower capacity usage. The model 01506-26 cartridge is compatible with the B-Pure, Nanopure II, old Nanopure, and Bantam systems, while model 01505-29 is compatible with E-Pure and quad-style Nanopure.
1 Year Warranty
Specifications
  • Capacity (grains)1700 as Calcium Carbonate
  • Compatible withAll NANOpure™ E-pure™, B-pure™, Bantam systems
  • FunctionHigh capacity, lower purity; use for making up stock solutions
  • DescriptionDeionization Cartridges, high capacity