Thermo Scientific 9512HPBNWP ISE electrode, Hi-Performance Ammonia, BNC
Representative image only.
Thermo Scientific 9512HPBNWP ISE electrode, Hi-Performance Ammonia, BNC
Thermo Scientific | Mfr: 9512HPBNWP| Item: 0572218
Designed to meet the demanding needs of both drinking water and wastewater applications
₹324,000.00INR / Each
₹324,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 9512HPBNWP ISE electrode, Hi-Performance Ammonia, BNC
Thermo Scientific 9512HPBNWP ISE electrode, Hi-Performance Ammonia, BNC
0572218
₹324,000.00INR / Each
+ =
Total Price 384200.00 ADD 2 ITEMS TO CART
Product Description
Includes
20 replacement membranes, preassembled electrode body with membrane, 60-mL of filling solution, and 1-m cable with BNC connector.
Key Features
  • Compliant with EPA testing method for ammonia measurement
  • Saves time to help make your lab more efficient
  • EPA-approved for ISE methods for analysis of wastewater
  • 1 year warranty
More About this Item
This Orion high performance ammonia electrode is extremely durable with a chemical-resistant translucent body. The easy-to-fill electrode comes marked with a fill line to avoid overfilling and allows you to monitor the fill solution level without disassembling the electrode. Membrane replacement options include cost-saving loose membranes or preassembled electrode body with membrane for the convenience of not having to install your own membrane.
1 Year Warranty
Specifications
  • ApplicationsHigh performance
  • Electrode TypeGas sensing
  • Electrode Body MaterialTranslucent plastic
  • Reference ElectrolyteAmmonia electrode filling solution 05722-22
  • ConnectorBNC
  • DescriptionISE electrode, Hi-Performance Ammonia, BNC