Thermo Scientific 951214 Membrane replace ammonia 20/box (Use with: 05722-18)
Representative image only.
Thermo Scientific 951214 Membrane replace ammonia 20/box (Use with: 05722-18)
Thermo Scientific | Mfr: 951214| Item: 0572224
Achieve optimum performance with fresh membrane
₹60,200.00INR / Box of 1
₹60,200.00INR / Box of 1
Buy It Together
Current Item
Thermo Scientific 951214 Membrane replace ammonia 20/box (Use with: 05722-18)
Thermo Scientific 951214 Membrane replace ammonia 20/box (Use with: 05722-18)
0572224
₹60,200.00INR / Box of 1
+ =
Total Price 60200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Replace old membranes on gas-sensing ion selective electrodes for accurate measurements.
1 Year Warranty
Specifications
  • Compatible WithAmmonia ISE electrode 05722-18
  • TypeLoose
  • DescriptionLoose Membrane, Ammonia; 20/Box