Thermo Scientific 7100-2444-078 Tube Adapter for 33851-25, 1"

Representative image only.

Thermo Scientific 7100-2444-078 Tube Adapter for 33851-25, 1"

Thermo Scientific | Mfr: 7100-2444-078| Item: 3385150

₹97,400.00INR / Each

₹97,400.00INR / Each

Buy It Together

Current Item

Thermo Scientific 7100-2444-078 Tube Adapter for 33851-25, 1"
Thermo Scientific 7100-2444-078 Tube Adapter for 33851-25, 1"
3385150

₹97,400.00INR / Each
+ + + =
Total Price 2168400.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Ceramic hearth plates, shelves and other parts and accessories for your muffle furnace.

1 Year Warranty
Specifications
  • DescriptionTube Adapter for 33851-25, 1"
  • Warranty1 Year
Reviews
Q&A