Texwipe TX757E Cleanroom swab; Cleanfoam tip and ESD handle; 0.15"L tip
Representative image only.
Texwipe TX757E Cleanroom swab; Cleanfoam tip and ESD handle; 0.15"L tip
Texwipe | Mfr: TX757E| Item: 3367774
Ideal for cleaning small surface areas
₹20,400.00INR / Bag of 500
₹20,400.00INR / Bag of 500
Buy It Together
Current Item
Texwipe TX757E Cleanroom swab; Cleanfoam tip and ESD handle; 0.15"L tip
Texwipe TX757E Cleanroom swab; Cleanfoam tip and ESD handle; 0.15"L tip
3367774
₹20,400.00INR / Bag of 500
+ =
Total Price 27500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Safely removes solvents and fluxes
  • Features low levels of nonvolatile residue
More About this Item
Foam swabs have sealed handles and are constructed from 100 pores per inch reticulated urethane foam — ideal for solvent dispersion and low-viscosity adhesives. Sealed foam swabs are perfect for removal of solvents and fluxes. Cleanfoam® swabs feature low levels of nonvolatile residue, particle generation, and are constructed of open-cell polyurethane foam for excellent solvent capacity.
Swabs with ESD handles eliminate damage from static electricity and are effective in all humidities.
Specifications
  • Tip Length (InchShort)5/32
  • ComplianceClass 100
  • Swab Total Length (InchShort)2 3/4
  • DescriptionCleanroom swab; Cleanfoam tip and ESD handle; 0.15"L tip