Texwipe TX708A Cleanroom swab; circular-foam tip and polypropylene handle; 0.81"L tip
Representative image only.
Texwipe TX708A Cleanroom swab; circular-foam tip and polypropylene handle; 0.81"L tip
Texwipe | Mfr: TX708A| Item: 3367756
Ideal for cleaning small surface areas
₹8,600.00INR / Bag of 100
₹8,600.00INR / Bag of 100
Buy It Together
Current Item
Texwipe TX708A Cleanroom swab; circular-foam tip and polypropylene handle; 0.81"L tip
Texwipe TX708A Cleanroom swab; circular-foam tip and polypropylene handle; 0.81"L tip
3367756
₹8,600.00INR / Bag of 100
+ =
Total Price 15700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Safely removes solvents and fluxes
  • Features low levels of nonvolatile residue
More About this Item
Foam swabs have sealed handles and are constructed from 100 pores per inch reticulated urethane foam — ideal for solvent dispersion and low-viscosity adhesives. Sealed foam swabs are perfect for removal of solvents and fluxes. Cleanfoam® swabs feature low levels of nonvolatile residue, particle generation, and are constructed of open-cell polyurethane foam for excellent solvent capacity.
Specifications
  • ComplianceClass 100
  • Tip Length (InchShort)13/16
  • Swab Total Length (InchShort)5 13/64
  • DescriptionCleanroom swab; circular-foam tip and polypropylene handle; 0.81"L tip