Texwipe TX700B Cleanroom swab; foam bud tip and hardwood handle; 1.10"L tip, 500/pk
Representative image only.
Texwipe TX700B Cleanroom swab; foam bud tip and hardwood handle; 1.10"L tip, 500/pk
Texwipe | Mfr: TX700B| Item: 3367603
Ideal for cleaning small surface areas
₹34,200.00INR / Bag of 500
₹34,200.00INR / Bag of 500
Buy It Together
Current Item
Texwipe TX700B Cleanroom swab; foam bud tip and hardwood handle; 1.10"L tip, 500/pk
Texwipe TX700B Cleanroom swab; foam bud tip and hardwood handle; 1.10"L tip, 500/pk
3367603
₹34,200.00INR / Bag of 500
+ =
Total Price 42600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Safely removes solvents and fluxes
  • Features low levels of nonvolatile residue
More About this Item
Foam swabs have sealed handles and are constructed from 100 pores per inch reticulated urethane foam — ideal for solvent dispersion and low-viscosity adhesives. Sealed foam swabs are perfect for removal of solvents and fluxes. Cleanfoam® swabs feature low levels of nonvolatile residue, particle generation, and are constructed of open-cell polyurethane foam for excellent solvent capacity.
Specifications
  • ComplianceNone
  • Tip Length (InchShort)1 7/64
  • Swab Total Length (InchShort)6 1/16
  • DescriptionCleanroom swab; foam bud tip and hardwood handle; 1.10"L tip, 500/pk