Testo 0602 0393 Temperature Probe, Type K, Surface
Representative image only.
Testo 0602 0393 Temperature Probe, Type K, Surface
Testo | Mfr: 0602 0393| Item: 3748618
Enhance your temperature monitoring
₹63,900.00INR / Each
for an additional
₹30,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹63,900.00INR / Each
Buy It Together
Current Item
Testo 0602 0393 Temperature Probe, Type K, Surface
Testo 0602 0393 Temperature Probe, Type K, Surface
3748618
₹63,900.00INR / Each
+ =
Total Price 106600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Fast response time (3 seconds) thanks to the thermocouple strip
  • Suitable for uneven surfaces
  • Measuring range short-term up to +500°C
  • 2 year warranty
More About this Item
These customized probes are designed to meet your specific needs in monitoring temperatures of food, liquid, and other samples.
Fast-action surface probe with sprung thermocouple strip, can be used on uneven surfaces, T/C Type K.
This probe has a 0.2" (5mm) shaft diameter, 0.5" (12mm) probe diameter, and 4.5" (115mm) shaft length.
Two year warranty
Specifications
  • DescriptionTemperature Probe, Type K, Surface