Testo 0563 0108 Waterproof Thermocouple Thermometer, -58 to 572F, Type T and K, IP65 Rated
Representative image only.
Testo 0563 0108 Waterproof Thermocouple Thermometer, -58 to 572F, Type T and K, IP65 Rated
Testo | Mfr: 0563 1080| Item: 9002524
Optimal choice for accuracy and maximum durability in food service applications
₹60,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹60,700.00INR / Each
Buy It Together
Current Item
Testo 0563 0108 Waterproof Thermocouple Thermometer, -58 to 572F, Type T and K, IP65 Rated
Testo 0563 0108 Waterproof Thermocouple Thermometer, -58 to 572F, Type T and K, IP65 Rated
9002524
₹60,700.00INR / Each
+ =
Total Price 105400.00 ADD 2 ITEMS TO CART
Product Description
Includes
standard immersion / penetration probe, soft case and 3 AAA batteries.
Key Features
  • Immersion / penetration probe for greater testing flexibility
  • Narrow probe tip for precision measurements
  • Conforms to HACCP and EN 13485
  • Rugged housing protection (IP67) for daily use and safe cleaning
  • Small and lightweight
  • 2 year warranty
More About this Item
Use the testo digital food thermometer, testo 108, for quick and convenient spot-checks on all types of food. Get instant feedback in seconds either in °C or °F on the large LCD display
from -58 to 572 °F. This model features a Type T or Type K probe depending upon your application range, Auto off and 0.9 °F accuracy. The meter is protected by a durable protective cover from water, dirt and impact.
Two year warranty
Specifications
  • Meter Measurement TypeThermocouple
  • Types Of Probes AcceptedType K, Type T
  • Min Temperature (° F)-58
  • Max Temperature (° F)572
  • Min Temperature (° C)-50
  • Max Temperature (° C)300
  • Resolution0.1°
  • IP RatingIP65
  • Display TypeLCD
  • Width (in)5 33/64
  • Depth (in)61/64
  • No. of Probe(s) Included1
  • Battery Life2500 hrs
  • Battery3 AAA (included)
  • DescriptionWaterproof Thermocouple Thermometer, -58 to 572F, Type T and KIP-65 Rated