Supco DVT4 Data View 4-Channel Temperature Data Logger with Display
Representative image only.
Supco DVT4 Data View 4-Channel Temperature Data Logger with Display
Supco | Mfr: DVT4| Item: 1800026
View temperature trends at a glance
₹98,100.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹98,100.00INR / Each
Buy It Together
Current Item
Supco DVT4 Data View 4-Channel Temperature Data Logger with Display
Supco DVT4 Data View 4-Channel Temperature Data Logger with Display
1800026
₹98,100.00INR / Each
+ =
Total Price 142800.00 ADD 2 ITEMS TO CART
Product Description
Includes
software, 6-ft (1.8-m) USB cable, and three AA batteries.
Key Features
  • Switch from text to graphical display for visual data monitoring
  • Compact design is ideal for any location
  • 1 year warranty
More About this Item
Units display real time temperature or temperature and humidity measurements, logging summary, sample rate, and memory left/used in text format. Measurement trends with high/low alarm set points are graphically displayed to easily monitor conditions. Data loggers are USB compatible allowing you to easily export data to text or Excel® files. Software is compatible with Windows® 98SE, ME, 2000, XP, and above.
Model 18002-26 instantly records four channels with included sensors (3 external and one internal). The large display shows all channels simultaneously, as well as temperature difference between the channels.
1 Year WarrantyDEW POINT
Specifications
  • Number of Channels4
  • Input TypeTemperature (three external, one internal)
  • Max Temperature (° F)160
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (° C)70
  • Temperature Resolution (° F)0.1
  • Temperature Accuracy (° F)±1
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±0.5
  • Memory SizeTotal data points: 87,040; Two temperatures: 43,420 points per channel; Three temperatures: 29,013 data points per channel; Four temperatures: 21,760 data points per channel
  • System RequirementsWindows® 98SE, ME, 2000, XP, Vista, 7. Compatible with 32 and 64 bit systems (not compatible with Windows NT or Windows 95); 5MB free disk space; 800 x 600 screen resolution; One free USB port
  • AlarmVisual over and under on all four channels
  • Battery Life1 year continuous use, 2 years average use
  • Battery Type3 AA (included)
  • Display TypeGraphic LCD
  • Length (in)4
  • Height (in)3
  • Width (in)1 1/2
  • DescriptionData View 4-Channel Temperature Data Logger with Display