Spill-X 76396 Caustic Spill Kit Agent Refill; 42 lb Pail

Representative image only.

Spill-X 76396 Caustic Spill Kit Agent Refill; 42 lb Pail

Spill-X | Mfr: 76396| Item: 8629352

Neutralizes spills for safer cleanup and handling

₹189,000.00INR / Each

₹189,000.00INR / Each

Buy It Together

Current Item

Spill-X 76396 Caustic Spill Kit Agent Refill; 42 lb Pail
Spill-X 76396 Caustic Spill Kit Agent Refill; 42 lb Pail
8629352

₹189,000.00INR / Each
+
Spill-X 78776 Acid Spill Treatment/Clean Up Kit
8629200

₹216,000.00INR / Each
=
Total Price 405000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Granular base reduces product waste and disposal costs
  • Speeds spill cleanup to limit worker exposure and minimize production downtime
  • Non-toxic, free-flowing dry chemical is environmentally friendly
  • Easy identification of agent with unique color
  • 1 year warranty

More About this Item

These refill agents are available in easy-to-store polypropylene containers. Order to replace specific or multipurpose spill treatment kit containers.

Economy-size pail of control agent covers a 3-gallon (11 L) spill. guide and instructions are included with each pail for start-to-finish cleanup.

1 Year Warranty
Specifications
  • Chemical TreatedCaustic
  • Agent ColorTan
  • ColorTan
  • DescriptionCaustic Spill Kit Agent Refill; 42 lb Pail
  • Warranty1 year
Documents & Certificates
Reviews
Q&A