Spill-X 78776 Acid Spill Treatment/Clean Up Kit
Representative image only.
Spill-X 78776 Acid Spill Treatment/Clean Up Kit
Spill-X | Mfr: 78776| Item: 8629200
Treat chemical spills in the lab or on the plant floor with a single step!
₹216,000.00INR / Each
₹216,000.00INR / Each
Buy It Together
Current Item
Spill-X 78776 Acid Spill Treatment/Clean Up Kit
Spill-X 78776 Acid Spill Treatment/Clean Up Kit
8629200
₹216,000.00INR / Each
+
Spill-X 77255 Acid Agent Refill, 6 containers
8629250
Call For Price
+ =
Total Price 216000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Spill Kit Treatment Guide, six polypropylene containers of agent, plastic case, one pair of rubber gloves and safety goggles, 100 pH test papers with color guide, scraper, two pick-up pans, and six disposal bags.
Key Features
  • Neutralizes hazardous liquid and chemical spills
  • Quick cleanups
  • 1 year warranty
More About this Item
These acid-neutralizing spill kits contain sorbents and other supplies which allow you to cleanup spills quickly. The easy-to-use shaker containers help neutralize hazardous liquid and chemical spills.
Order agent refills separately.
Spill-X-A® Acid Neutralizer neutralizes and solidifies most organic and inorganic acids, including hydrofluoric acid.
1 Year Warranty
Specifications
  • Chemical TreatedAcid
  • Agent ColorRed
  • DescriptionAcid Spill Treatment/Clean Up Kit