Shimpo DT-207LR Contact/Noncontact LCD Tachometer with 6" Circ. Measuring Wheel
Representative image only.
Shimpo DT-207LR Contact/Noncontact LCD Tachometer with 6" Circ. Measuring Wheel
Shimpo | Mfr: DT-207LR: TACHOMETER| Item: 9880631
Check out these features!
₹107,000.00INR / Each
for an additional
₹39,200.00INR / Each
for an additional
₹37,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹107,000.00INR / Each
Buy It Together
Current Item
Shimpo DT-207LR Contact/Noncontact LCD Tachometer with 6" Circ. Measuring Wheel
Shimpo DT-207LR Contact/Noncontact LCD Tachometer with 6" Circ. Measuring Wheel
9880631
₹107,000.00INR / Each
+
Shimpo CONE Accessory Convex tip for tachometers
0820370
₹2,300.00INR / Each
+
InnoCal NIST-Traceable Calibration; Tachometer
1705000
₹37,400.00INR / Each
=
Total Price 146700.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Sophisticated laser beam detection
  • Exceptional accuracy
  • Length and rate functions offer expanded measurement capabilities
  • Multi-mode speed selection
  • Broad speed range
  • Hardy aluminum construction, assures a maintenance-free performance for many years
  • Large 5 digit display
  • Expansive 10 test memory capacity
  • Stores last / min / max readings for thorough equipment / process inspection
  • Advanced laser beam detection enables safe RPM measurement from 20 feet
  • Extremely accurate
  • All metal construction
  • Includes a standard calibrated 6'' circumference wheel with locking screw for contact rate and length applications
  • 1 year warranty
More About this Item
A laser allows precise RPM measurement from 20 feet! Our new tachometers incorporate laser technology yet retain rugged construction and reliable quality components; both offer incredible value and versatility as they perform non-contact and contact applications via the included contact adapter. Capture fast speeds with laser precision Incorporating the precise laser technology and user-friendly functions as our standard tachometers, the Deluxe Models provide exceptional accuracy in measuring speed and length via a 12'' wheel; the larger wheel rotates more slowly than a standard 6'' wheel, enabling reliable and safe registration of high speeds. These superior instruments also feature a contact adapter, conveniently allowing both non-contact and contact measurement.
1 Year WarrantyConformite Europeenne
Specifications
  • Max Measuring Distance (in)20
  • Display Type5-digit LCD
  • MemoryLast 10 readings, min/max
  • DescriptionContact/Noncontact LCD Tachometer with 6" Circ. Measuring Wheel