InnoCal A2LA ISO17025 Accredited Calibration; Tachometer
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Tachometer
InnoCal | Item: 1750031
₹39,200.00INR / Each
₹39,200.00INR / Each
Buy It Together
Current Item
InnoCal A2LA ISO17025 Accredited Calibration; Tachometer
InnoCal A2LA ISO17025 Accredited Calibration; Tachometer
1750031
₹39,200.00INR / Each
+
Traceable Noncontact Tachometer with Red LED
9876708
₹21,800.00INR / Each
=
Total Price 61000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • A2LA Accredited Calibration. View our scope of accreditation
  • Calibration service for contact tachometers (mechanical), non-contact tachometers (optical), and combination tachometers featuring both contact and non-contact options
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005
Specifications
  • Calibration TypeA2LA ISO17025 Accredited
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test PointsMechanical: 10 Points; Optical: 12 test point
  • Calibration Test PointsThroughout range of instrument
  • Calibration Test RangeMechanical: 60 to 24,000 rpm; Optical: 0 to 200,000 rpm
  • DescriptionA2LA ISO17025 Accredited Calibration; Tachometer