Sartorius Arium® Smart Station Ultrapure Water Remote Dispenser, Benchtop

Representative image only.

Sartorius Arium® Smart Station Ultrapure Water Remote Dispenser, Benchtop

Sartorius | Mfr: H2O-ARST-UP-T| Item: 9929393

Remotely dispense pure and ultrapure water from water purification system or Bagtank

Call For Price

Call For Price

Buy It Together

Current Item

Sartorius Arium® Smart Station Ultrapure Water Remote Dispenser, Benchtop
Sartorius Arium® Smart Station Ultrapure Water Remote Dispenser, Benchtop
9929393

Call For Price
+ + =
Total Price 0 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Accurate volume dispensing for fast, precise buffer and sample preparation
  • Compact design easily fits into any lab environment
  • Easy-to-navigate touch-activated color display
  • Stepless height adjustment to fill different size containers
  • Adjust flow rate from drop-by-drop to 2 L/min
  • Compatible with Arium Pro and Arium Comfort water systems
  • Compact bentop unit takes up minimal space
  • 2 year warranty

More About this Item

Combine the Arium smart station with a water purification system or a Bagtank to remotely dispense pure or ultrapure water. These compact smart stations take up minimal space for easy and space-saving integration in your lab work area.

Use the large touch-activated color display to access all dispense options and view important quality parameters. Flexible, stepless height adjustment and a full 360° rotating dispenser arm make it easy to dispense water into a broad range of containers. The ergonomic design of the dispenser arm supports left- and right-hand operation. Adjust dispense volume from drop-by-drop to 2 L/min to meet your buffer and sample preparation needs.

Connect to Arium Pro and Arium Comfort water systems to dispense ultrapure water at the point of use.

Conformite Europeenne
Specifications
  • Compatible WithArium® Pro and Arium Comfort
  • Height (in)13.5
  • Width (InchShort)6 3/4
  • Depth (InchShort)6 3/16
  • Width (cm)17.2
  • Height (cm)34.3
  • Depth (cm)15.7
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Power (amps)2.5
  • DescriptionBenchtop Ultrapure Water Remote Dispenser
  • Warranty2 years
Reviews
Q&A