Sartorius Arium® Pure Water Tubing Extension Set
Representative image only.
Sartorius Arium® Pure Water Tubing Extension Set
Sartorius | Mfr: H2O-ATES-P| Item: 9929394
Keep your Arium water purification system running smoothly
Call For Price
Call For Price
Buy It Together
Current Item
Sartorius Arium® Pure Water Tubing Extension Set
Sartorius Arium® Pure Water Tubing Extension Set
9929394
Call For Price
+
Sartorius Arium Mini Water Purification System
9929310
Call For Price
+
Sartorius Arium® Pro Basic Wall-Mount System
9929340
Call For Price
+ +
Sartorius Arium® Mini Plus Starter Set
9929444
Call For Price
+ =
Total Price 0 ADD 6 ITEMS TO CART
Product Description
Key Features
  • Use with the Arium Smart Station pure water remote dispenser
  • Extend the operating range from 2 to 4 meters
  • 2 year warranty
More About this Item
Filters, cartridges, and other accessories for water purification.
The tubing extension set makes a fast and reliable connection between the Arium smart station and a pure water system or bagtank.
Specifications
  • Material1/4" Polyethylene (PVDF/PE) tubing
  • Compatible WithArium Smart Station pure water remote dispenser
  • ConnectionsEthernet CAT6A
  • Length (InchShort)157 1/2
  • Length (cm)400
  • DescriptionPure Water Tubing Extension Set for Smart Station