Sartorius Arium® Conversion Kit
Representative image only.
Sartorius Arium® Conversion Kit
Sartorius | Mfr: H2O-ACK-D| Item: 9929418
Keep your Arium water purification system running smoothly
Call For Price
Call For Price
Buy It Together
Current Item
Sartorius Arium® Conversion Kit
Sartorius Arium® Conversion Kit
9929418
Call For Price
+
Sartorius Arium Mini Water Purification System
9929310
Call For Price
+
Sartorius Arium® Pro Basic Wall-Mount System
9929340
Call For Price
+ +
Sartorius Arium® Mini Plus Starter Set
9929444
Call For Price
+ =
Total Price 0 ADD 6 ITEMS TO CART
Product Description
Key Features
  • Use with Arium Comfort or Pro benchtop water systems
  • Convert your benchtop system into a built-in version
  • 1 year warranty
More About this Item
Filters, cartridges, and other accessories for water purification.
The conversion kit helps provide optimal integration of your Arium water systems into your laboratory. Through variable wall installation of the display and dispenser unit and extention of tubing, the kit helps create more space on and above the laboratory bench.
Specifications
  • MaterialPVDF
  • Compatible WithArium Comfort or Pro benchtop
  • DescriptionConversion Kit for Below Bench Mounting or Remote Displays