Rex Gauge TEST BLOCK KIT TYPE A Test Block Kit for Type A Durometers; 7 Blocks Covering Range 30 to 90
Representative image only.
Rex Gauge TEST BLOCK KIT TYPE A Test Block Kit for Type A Durometers; 7 Blocks Covering Range 30 to 90
Rex Gauge | Mfr: TEST BLOCK KIT TYPE A| Item: 5979250
Maintain accurate durometer readings
₹74,700.00INR / Each
₹74,700.00INR / Each
Buy It Together
Current Item
Rex Gauge TEST BLOCK KIT TYPE A Test Block Kit for Type A Durometers; 7 Blocks Covering Range 30 to 90
Rex Gauge TEST BLOCK KIT TYPE A Test Block Kit for Type A Durometers; 7 Blocks Covering Range 30 to 90
5979250
₹74,700.00INR / Each
+ =
Total Price 89400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Values ranging from 30 to 90 durometer
  • 1 year warranty
More About this Item
The kit acts as a quick reference check for the status of your durometer and lets you develop the proper feel for use of the gauge. Includes seven color-coded rubber test blocks of various type A hardness values ranging from 30 to 90 durometer.
1 Year Warranty
Specifications
  • DescriptionTest Block Kit for Type A Durometers; 7 Blocks Covering Range 30 to 90