Rex Gauge TB-1-Navy Test Block for Durometers - Type-A, Single Block, Color Navy
Representative image only.
Rex Gauge TB-1-Navy Test Block for Durometers - Type-A, Single Block, Color Navy
Rex Gauge | Mfr: SINGLE TEST BLACK A - TB-1-NAVY| Item: 5979254
Optimize performance of your Rex Gauge durometer kit test block
₹14,700.00INR / Each
₹14,700.00INR / Each
Product Description
Key Features
  • Keep your durometer kit test block running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Rex Gauge durometer kit test block.
This shore A test block aids in obtaining accurate readings from your durometer as it allows you to develop the proper "feel" for the gauge. Rex Gauge's test blocks give you the ability to develop the proper feel, and, while it is not to be used for calibration purposes, it enables the user to perform a quick check for proper durometer operation.
1 Year Warranty
Specifications
  • DescriptionShore A Test Block for Durometers