ResinTech VP-17-4030-HN VP Series Color Changing High Purity Mixed Bed, Hose Nipple
Representative image only.
ResinTech VP-17-4030-HN VP Series Color Changing High Purity Mixed Bed, Hose Nipple
ResinTech | Mfr: VP-17-4030-HN| Item: 9928487
Reduce operating costs while maintaining peak performance
₹47,600.00INR / Each
₹47,600.00INR / Each
Buy It Together
Current Item
ResinTech VP-17-4030-HN VP Series Color Changing High Purity Mixed Bed, Hose Nipple
ResinTech VP-17-4030-HN VP Series Color Changing High Purity Mixed Bed, Hose Nipple
9928487
₹47,600.00INR / Each
+ + +
ResinTech IX Series High Purity Color Changing DI
9928506
₹35,000.00INR / Each
+ =
Total Price 192800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Activated carbon mixed bed, color-changing Hose barb, 3/8"
  • 1 year warranty
More About this Item
Use these filtration cartridges in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.
1 Year Warranty
Specifications
  • MediaHigh Purity, Mixed Bed, Color Changing
  • Capacity1360 grains
  • Capacity (grains)1360
  • Connection2 x 3/8" Hose Barb
  • Compatible withARIES, Unix, Barnstead, Vaponics
  • Max Inlet Pressure (PSI)30
  • Max Flow Rate (LPM)3.8
  • Max Flow Rate (GPH)60
  • Diameter (in)3.5
  • Length (in)17.8
  • DescriptionFilter Cartridge, High Purity Mixed Bed, Color Changing Resin, 3/8" Hose Barb