ResinTech VP-17-1050-HN VP Series Organics Removal, Hose Nipple
Representative image only.
ResinTech VP-17-1050-HN VP Series Organics Removal, Hose Nipple
ResinTech | Mfr: VP-17-1050-HN| Item: 9928489
Reduce operating costs while maintaining peak performance
₹35,900.00INR / Each
₹35,900.00INR / Each
Buy It Together
Current Item
ResinTech VP-17-1050-HN VP Series Organics Removal, Hose Nipple
ResinTech VP-17-1050-HN VP Series Organics Removal, Hose Nipple
9928489
₹35,900.00INR / Each
+ + +
ResinTech IX Series High Purity Color Changing DI
9928506
₹35,000.00INR / Each
+ =
Total Price 181100.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Activated carbon Hose barb, 3/8"
  • 1 year warranty
More About this Item
Use these filtration cartridges in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.
1 Year Warranty
Specifications
  • MediaActivated Carbon
  • Capacity2000 gal.
  • Capacity (Gal)2000
  • Connection2 x 3/8" Hose Barb
  • Compatible withARIES, Unix, Barnstead, Vaponics
  • Max Inlet Pressure (PSI)30
  • Max Flow Rate (LPM)3.8
  • Max Flow Rate (GPH)60
  • Diameter (in)3.5
  • Length (in)17.8
  • DescriptionFilter Cartridge, Activated Carbon Resin, 3/8" Hose Barb