ResinTech VP-17-0001 VP Series Sanitization Cartridge
Representative image only.
ResinTech VP-17-0001 VP Series Sanitization Cartridge
ResinTech | Mfr: VP-17-0001| Item: 9928530
Reduce operating costs while maintaining peak performance
₹44,700.00INR / Each
₹44,700.00INR / Each
Buy It Together
Current Item
ResinTech VP-17-0001 VP Series Sanitization Cartridge
ResinTech VP-17-0001 VP Series Sanitization Cartridge
9928530
₹44,700.00INR / Each
+
ResinTech IX Series High Purity Color Changing DI
9928506
₹35,000.00INR / Each
+ + =
Total Price 137900.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • 1 year warranty
More About this Item
Use these filtration cartridges in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.
1 Year Warranty
Specifications
  • DescriptionSanitization Cartridge