Qsonica 4420 Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia
Representative image only.
Qsonica 4420 Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia
Qsonica | Mfr: 4420| Item: 0471192
Microtip Probes for use with Cole-Parmer and Qsonica Q500 and Q700 Sonicators
₹64,500.00INR / Each
₹64,500.00INR / Each
Buy It Together
Current Item
Qsonica 4420 Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia
Qsonica 4420 Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia
0471192
₹64,500.00INR / Each
+ +
Cole-Parmer Coupler for 500- and 750-Watt Processors
0471085
₹56,600.00INR / Each
=
Total Price 121100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.
Two year warranty
Specifications
  • Max Sample Size (mL)50
  • Tip Diameter (in)0.25
  • Tip Diameter (cm)0.635
  • DescriptionUltrasonic Cell Disruptor Microtip Probes, 1/4" dia