Pyrex 7079-2N Disposable Glass Serological Pipette, 2 mL, Bulk, 700/Cs

Representative image only.

Pyrex 7079-2N Disposable Glass Serological Pipette, 2 mL, Bulk, 700/Cs

Pyrex | Mfr: 7079-2N| Item: 3456825

Disposable/Reusable glass serological non-sterile, non-plugged pipettes

Call For Price

Call For Price

Buy It Together

Current Item

Pyrex 7079-2N Disposable Glass Serological Pipette, 2 mL, Bulk, 700/Cs
Pyrex 7079-2N Disposable Glass Serological Pipette, 2 mL, Bulk, 700/Cs
3456825

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Pipettes do not have plugged-ends and come bulk-packed
  • Negative graduations for versatiity
  • Reference: ASTM E-714

More About this Item

These non-sterile pipettes are calibrated "to deliver" (TD) with blow out. Slender, fire-polished tips allow for fast and easy pipetting with burr-free, even flow rates.
NOTE: Pipetting by mouth can be hazardous to your health. We suggest using a mechanical pipetting device.

Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)2000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)2
  • Max Dispensing (mL)2
  • Graduations (mL)0.01
  • Overall Length (InchShort)11.41
  • Overall Length (mm)290
  • Tip StyleStandard
  • Body MaterialGlass
  • CalibrationTo Deliver (TD)
  • DescriptionDisposableGlassSerologicalPipette,2mL,Bulk,700/Cs
Reviews
Q&A