Pyrex 2116-LSO 2116 Economical Glass Burette Replacement Stopcock; 1/Cs
Representative image only.
Pyrex 2116-LSO 2116 Economical Glass Burette Replacement Stopcock; 1/Cs
Pyrex | Mfr: 2116-LSO| Item: 3454210
Replacement components reduce cost
₹21,700.00INR / Case of 1
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹21,700.00INR / Case of 1
Buy It Together
Current Item
Pyrex 2116-LSO 2116 Economical Glass Burette Replacement Stopcock; 1/Cs
Pyrex 2116-LSO 2116 Economical Glass Burette Replacement Stopcock; 1/Cs
3454210
₹21,700.00INR / Case of 1
+ +
Cole-Parmer Acrylic Burette, 25 mL; 1/Pk
3454051
₹9,900.00INR / Each
+
Cole-Parmer Burette, 10 mL With Glass Stopcock
6310051
₹12,100.00INR / Each
=
Total Price 58200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from burette (buret) or replacement components
  • Replacement stopcock for 34542-00 and -02 burettes
More About this Item
Burette (buret) components are designed to be replaced eliminating the need to purchase an entirely new burette if any part is damaged. Each burette has tooled top and bottom, white enamel graduations, and replaceable screw-thread locking PTFE stopcock. Replaceable tip end is ground to prevent tip from falling out.
Specifications
  • Capacity (mL)25
  • Tolerance0.1
  • Description2116 Economical Glass Burette Replacement Stopcock; 1/Cs