Cole-Parmer® Class B Burettes (Burets)
No more difficult readings
  • Easy-to-read burette (buret) with imprinted red enamel graduations
Burettes (burets) are manufactured from high quality boresilicate tubing to provide a high degree of accuracy, reliablity, and durability. Burettes have a glass stopcock and imprinted red enamel graduations.
12,100.00 - ₹69,400.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Tolerance
Availability Pricing
COMPARE
10 0.05 0.02
₹12,100.00INR / Each
COMPARE
25 0.1 0.05
₹12,100.00INR / Each
COMPARE
50 0.1 0.05
₹13,000.00INR / Each
COMPARE
100 0.2 0.2
₹17,600.00INR / Each
COMPARE
25 0.1 0.05
₹55,700.00INR / Case of 6
COMPARE
50 0.1 0.05
₹69,400.00INR / Case of 6
COMPARE
100 0.2 0.2
₹54,200.00INR / Case of 4
Showing 1- 7 of 7