Powerblanket GHT2002J-FS Digital Thermostatic Controller
Representative image only.
Powerblanket GHT2002J-FS Digital Thermostatic Controller
Powerblanket | Mfr: GHT2002JFS| Item: 4202515
Use with any Powerblanket heater
₹71,100.00INR / Each
₹71,100.00INR / Each
Buy It Together
Current Item
Powerblanket GHT2002J-FS Digital Thermostatic Controller
Powerblanket GHT2002J-FS Digital Thermostatic Controller
4202515
₹71,100.00INR / Each
+ + =
Total Price 418600.00 ADD 3 ITEMS TO CART
Product Description
Includes
6-ft (1.8-m) input power supply cord with plug, 6-ft (1.8-m) temperature sensor lead with steel encased temperature probe, hanger kit (installed), and 2-ft (0.6-m) output cord with connector/receptacle.
Key Features
  • Adjust temperature from ambient to the heater's maximum
  • Watertight—NEMA 4X, rugged, steel-enclosed casing
  • External probe included for automatic temperature adjustments
  • 1 year warranty
More About this Item
Plug the temperature controller into a GFCI protected power supply and plug your Powerblanket heating product into the receptacle on the controller—it's that easy! The controller will power up when plugged in. Use the steel-tipped temperature sensor to monitor temperature between the container and heater—controller will adjust the output to maintain the set temperature.
Do not submerge the temperature sensor—it is not waterproof.
1 Year Warranty
Specifications
  • Max Temperature (° F)212
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (DegreeC)100
  • Resolution
  • Data LoggingNo
  • Enclosure RatingNEMA 4X
  • Display TypeDigital
  • Power (VAC)110 - 120
  • DescriptionDigital Thermostatic Controller