BriskHeat® SDC Benchtop On/Off Temperature Controllers with Digital Display
Easy-to-use programmable digital controller delivers plug and play design
  • Benchtop On/Off digital temperature controller for simple heating protocols
  • Easy-to-read measurements with 0.5” high, 3-digit, red LED display
  • Enhanced safety with audible and visual alarm indicating error
These SDC-series digital on/off temperature controllers provide basic control and connect easily to heating tape without any additional wiring. The large display is easy to read and simple to program alarms, set-points, timer, and much more.
Compatible with a wide range of heaters, these controllers include a factory-installed thermocouple sensor. They are benchtop-friendly with a compact 5.5” x 4.25” x 1.75” profile. All controllers feature an adjustable hysteresis setting from 1° to 99°.
Choose from °F or °C displays; heaters are sold separately.
82,500.00 - ₹83,600.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
BriskHeat SDC120JF-A SDC Benchtop Temperature Controller with Digital Display, Type J , °F display; 120 VAC 3612099 Mfr # SDC120JF-A
COMPARE
120 Benchtop Temperature Controller with Digital Display, Type J , °F display; 120 VAC
₹82,500.00INR / Each
BriskHeat SDC120JC-A SDC Benchtop Temperature Controller with Digital Display, Type J , °C display; 120 VAC 3612100 Mfr # SDC120JC-A
COMPARE
120 Benchtop Temperature Controller with Digital Display, Type J , °C display; 120 VAC
₹82,500.00INR / Each
BriskHeat SDC120KF-A SDC Benchtop Temperature Controller with Digital Display, Type K , °F display; 120 VAC 3612101 Mfr # SDC120KF-A
COMPARE
120 Benchtop Temperature Controller with Digital Display, Type K , °F display; 120 VAC
₹82,500.00INR / Each
BriskHeat SDC120KC-A SDC Benchtop Temperature Controller with Digital Display, Type K , °C display; 120 VAC 3612102 Mfr # SDC120KC-A
COMPARE
120 Benchtop Temperature Controller with Digital Display, Type K , °C display; 120 VAC
₹82,500.00INR / Each
BriskHeat SDC240JF-A SDC Benchtop Temperature Controller with Digital Display, Type J , °F display; 240 VAC 3612103 Mfr # SDC240JF-A
COMPARE
240 Benchtop Temperature Controller with Digital Display, Type J , °F display; 240 VAC
₹82,500.00INR / Each
BriskHeat SDC240JC-A SDC Benchtop Temperature Controller with Digital Display, Type J , °C display; 240 VAC 3612104 Mfr # SDC240JC-A
COMPARE
240 Benchtop Temperature Controller with Digital Display, Type J , °C display; 240 VAC
₹82,500.00INR / Each
BriskHeat SDC240KC-A SDC Benchtop Temperature Controller with Digital Display, Type K , °C display; 240 VAC 3612106 Mfr # SDC240KC-A
COMPARE
240 Benchtop Temperature Controller with Digital Display, Type K , °C display; 240 VAC
₹83,600.00INR / Each
Showing 1- 7 of 7