Partlow 10" Circular Chart Recorder Paper, 0 to 20, 7 Days, 100/Box

Representative image only.

Partlow 10" Circular Chart Recorder Paper, 0 to 20, 7 Days, 100/Box

Partlow | Mfr: 00214755| Item: 4900307

10 or 12" circular replacement chart recorder paper

₹11,400.00INR / Each

₹11,400.00INR / Each

Buy It Together

Current Item

Partlow 10" Circular Chart Recorder Paper, 0 to 20, 7 Days, 100/Box
Partlow 10" Circular Chart Recorder Paper, 0 to 20, 7 Days, 100/Box
4900307

₹11,400.00INR / Each
+ =
Total Price 913400.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • A variety of ranges and speeds
  • Fits Partlow MRC 7000

More About this Item

Choose the paper size and type compatible with your application and Partlow chart recorder.

Specifications
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)10
  • Description10" Circular Chart Recorder Paper, 0 to 20, 7 Days, 100/Box
Reviews
Q&A