Environmental Express Oakton pHTestr® 10 Waterproof BNC Pocket pH Tester

Representative image only.

Environmental Express Oakton pHTestr® 10 Waterproof BNC Pocket pH Tester

Customizable to your application – use with any pH electrode with a BNC connector

₹23,300.00INR / Each

for an additional
₹18,700.00INR / Each
for an additional
₹25,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹23,300.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton pHTestr® 10 Waterproof BNC Pocket pH Tester
Environmental Express Oakton pHTestr® 10 Waterproof BNC Pocket pH Tester
3563414

₹23,300.00INR / Each
+ +
InnoCal NIST-Traceable Calibration; pH Tester or Pen
1710145

₹18,700.00INR / Each
+ + + + + =
Total Price 206100.00 ADD 8 ITEMS TO CART
Product Description

Includes

protective plastic storage case, lanyard, and 4 A76 batteries.

Key Features

  • Versatility of a handheld meter at a pocket price
  • IP67-rated protects meter from water, dust, and shock while offering chemical resistance
  • Meter floats—accidental drops into water will not result in lost or damaged meters
  • 1 year warranty

More About this Item

Simply choose the electrode best suited for your application, connect the pHTestr BNC 10, calibrate, and measure. Change electrodes to suit your application without having to purchase another meter. Order electrodes separately.

Large display and tactile keypad allow for easy calibration along with USA and NIST auto-buffer recognition. Hold function freezes reading on the display for easy recording. Additional features include Valox® sensor housing, self-diagnostic error messages, a battery-life indicator, and an auto-off function to maximize battery life.

1 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier
Specifications
  • Meter TypeTester
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 8-pin mini-DIN
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Min Temperature (° F)23
  • Max Temperature (° F)122
  • Temperature compensationManual
  • WaterproofYes
  • Battery4 A76 (included)
  • Diameter (cm)3.81
  • Diameter (in)1 1/2
  • Description10 Waterproof BNC Pocket pH Tester
  • Warranty1 year
Reviews
Q&A