Environmental Express Oakton PCTSTestr™ 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester

Representative image only.

Environmental Express Oakton PCTSTestr™ 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester

Premium multi parameter pocket testers for field and lab testing

₹69,000.00INR / Each

for an additional
₹18,700.00INR / Each
for an additional
₹25,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹69,000.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton PCTSTestr™ 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester
Environmental Express Oakton PCTSTestr™ 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester
3563435

₹69,000.00INR / Each
+ + +
InnoCal NIST-Traceable Calibration; pH Tester or Pen
1710145

₹18,700.00INR / Each
+ + +
Environmental Express Buffer Solution, pH 10.01; 500 mL
0065408

₹2,100.00INR / Pint of 1
+ + =
Total Price 173600.00 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Available in pH, Conductivity, TDS, Salinity, and ORP models
  • Replaceable, double-junction sensors for durability and longevity against aggressive samples
  • Four-button keypad for technician level use
  • Leakproof cap functions as a sample cup, hydrating cup, and stabilizing base to stand the meter upright
  • Automatic temperature compensation ensures accurate reading in varying environments

More About this Item

Our most advanced line of pocket testers, the Oakton 50 Series provides single and multimeter options for measuring pH, Conductivity, TDS, Salinity, and ORP. Replaceable double junction sensors provide durability and longevity against highly caustic, volatile or hazardous samples in both lab and field applications. Waterproof and designed to float in water, they are ideal for field-based testing environments – with a convenient loop for security line attachment.

50 series pocket meters include a replaceable double-junction electrode sensor that is designed for more aggressive applications, including wastewater and solutions containing sulfides, heavy metals, and TRIS. The electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage and allow for easy maintenance and replacement. A Valox sensor housing resists most chemicals.

An easy-to-use four-button keypad allows for technician level use and measurement with quick access to a variety of functions. Up to 3 calibration points with auto-buffer recognition at both USA and NIST standard buffers. Additional features include an automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.


Oakton PCTSTestr™ 50 is the ultimate pocket multimeter for pH, conductivity, TDS, salinity, and temperature! Great for water, wastewater, laboratory or plant use, these multiparameter testers feature long-life pH electrodes with a wide range of sample compatibility. Pin-style conductivity sensors feature stainless steel electrodes for chemical resistance and durability. A replaceable sensor module saves you time and money versus replacing the entire meter.

These testers bring the performance of your lab instruments out into the field. Advanced features of the PCTSTestr 50 include Conductivity, TDS, and Salinity auto-ranging based on the concentration of your sample, as well as up to 3-point pH calibration with NIST or USA auto buffer recognition. An auto-shutoff, user-adjustable temperature coefficient, and low-battery indicator round-out the robust functionality of this pocket tester.

1 Year WarrantyConformite EuropeenneIP 67ISO 9001:2008 Certified Supplier
Specifications
  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)-1
  • Max pH (pH)15
  • pH Resolution0.01
  • pH Accuracy±0.01
  • Buffer RecognitionUSA 4.01, 7.00, 10.01 and NIST 4.01, 6.86, 9.18
  • CalibrationUp to 3 points
  • Min Conductivity0 µS
  • Max Conductivity20 mS
  • Conductivity Accuracy±1% full scale
  • Min TDS0 ppm
  • Max TDS10.0 ppt
  • TDS Resolution10 ppt
  • TDS Accuracy+/- 1% FS
  • Min Salinity0.00 ppt
  • Max Salinity10.0 ppt
  • Salinity Resolution0.01 ppt
  • Salinity Accuracy+/- 1% FS
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Min Temperature (° F)32
  • Max Temperature (° F)140
  • Temperature Resolution0.1°C / 0.1°F
  • Temperature Accuracy0°C to 50°C (±0.5°C / ±0.9°F ); from 50°C to 60°C (±1.0°C / ±1.8 °F )
  • Temperature compensationYes (Automatic Temperature Compensation)
  • WaterproofYes
  • Battery4 AAA (included)
  • Battery life (hr)150
  • DescriptionWaterproof Pocket pH/Cond/TDS/Salinity Tester, Premium 50 Series
  • Warranty1 year
Reviews
Q&A