Environmental Express Oakton EcoTestr™ pH 2+ Pocket pH Meter

Representative image only.

Environmental Express Oakton EcoTestr™ pH 2+ Pocket pH Meter

Economical, field-ready testers that are easy to use

₹29,400.00INR / Each

for an additional
₹18,700.00INR / Each
for an additional
₹25,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹29,400.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton EcoTestr™ pH 2+ Pocket pH Meter
Environmental Express Oakton EcoTestr™ pH 2+ Pocket pH Meter
3542301

₹29,400.00INR / Each
+ + +
InnoCal NIST-Traceable Calibration; pH Tester or Pen
1710145

₹18,700.00INR / Each
+
Cole-Parmer Digital Counter Pen
2061030

₹4,000.00INR / Each
+ + + + + =
Total Price 356200.00 ADD 10 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Large, easy-to-read LCD screen
  • User-friendly, two-button interface
  • Rugged IP67-rated housing is waterproof, dustproof, and floats in water

More About this Item

Providing value without compromise, Oakton® EcoTestr™ 1 Series waterproof testers are easy to use and read. The large, dual-line LCD provides measurement and temperature information, as well as current battery life. The two-button interface allows for simple navigation of the meter menus and quick sample measurement. Designed to float in water, they are ideal for field testing applications such as lakes and streams – you can even attach a line to the housing for easy retrieval.

Additional features include a long-lasting durable electrode with polymer reference electrolyte; status icons for measurement stability, calibration, and battery life; and self-diagnostics. The leakproof cap with fill line functions as a sample/calibration and hydrating cup. The tester, powered by four AAA batteries, can operate for up to 400 hours continuously. The unit will auto power-off after 8 minutes of nonuse to conserve power.

The EcoTestr pH 2+ has a four-key interface which provides simplified menu navigation and convenient one-touch calibration. The LCD display is large with indicators for battery life, readiness, and calibration. It also shows both the parameter and temperature readings at the same time.

Ideal for field use, the EcoTestr pH 2+ comes with a leakproof cap that can be attached to the top of the meter when not being used—no more lost caps! The cap features a fill line allowing it to serve as a sample cup with proper measurement for accurate readings. A wide base helps to keep the meter upright for hands-free measurements.

The IP67-rated waterproof housing is compatible with lanyards to prevent loss or dropping but just in case, it also floats! The EcoTestr pH 2+ is suitable for a variety of applications, including environmental monitoring, water treatment, agriculture, and pools and spas.

1 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeTester
  • Display typeDigital
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.1 pH
  • pH Accuracy±0.1 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Temperature Resolution0.1°F (0.1°C)
  • Temperature Accuracy±0.9°F (0.5°C) + 1 LSD
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 A76 (included)
  • Battery life (hr)100
  • DescriptionPocket pH Meter
  • Warranty1 year
Reviews
Q&A