Nederman Wall Bracket for 33700 Series Extractor Arms
Representative image only.
Nederman Wall Bracket for 33700 Series Extractor Arms
Nederman | Mfr: 10550635| Item: 3370090
Up to 650 cfm air capacity for particles, smoke, and dust
₹44,000.00INR / Each
₹44,000.00INR / Each
Buy It Together
Current Item
Nederman Wall Bracket for 33700 Series Extractor Arms
Nederman Wall Bracket for 33700 Series Extractor Arms
3370090
₹44,000.00INR / Each
+
Cole-Parmer Ducted Wall Canopy Fume Hood, 36" W
3373130
₹951,000.00INR / Each
+ =
Total Price 1390000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 3 year warranty
More About this Item
Fume extractor arms are perfect for welding booths and other industrial applications requiring large air capacities. Arms come in four lengths: 7 ft; 10 ft; 14 ft; and telescopic, which measures 34" when retracted, 5 ft when fully extended. Telescopic and 7-ft arms are suitable for small work areas; 10-ft and 14-ft arms are best for wider work areas. Specially designed friction joints allow hoods to be positioned at any point within the work area. Arms are supported by a 360deg swivel elbow for unlimited maneuverability. Maximum operating temperature is 158degF (70degC). Assembly is required for all models.
The 7-ft, 10-ft, and 14-ft arms include 10" x 12" oval polycarbonate hood with handle and damper, and 6-1/4" OD PVC fiberglass-reinforced hose. Telescopic a
3 Year Warranty
Specifications
  • DescriptionWall Bracket for 33700 Series Extractor Arms
  • Warranty3 Years