Cole-Parmer® Ducted Canopy Hoods
Capture and eliminate contaminated air at the source
  • Choose from wall, island, or corner styles
  • Easily incorporate into existing labs and classrooms
Constructed of chemical-resistant composite materials, these molded, one-piece canopy hoods are lightweight and designed to collect and exhaust corrosive vapors, heat, steam, and odors when mounted over areas with baths and circulators, small mixers, hot plates, or other equipment. If your application requires hood material of 304 stainless steel, contact our Application Specialists.
To customize your canopy hood, order clear fiberglass side panels separately below. Side panels attach directly to side of hood and help to contain fumes and minimize effects of cross drafts. Vapor-proof incandescent light fixture 33731-88 adds additional light to your working area and mounts easily to the inside of your canopy.
Wall and corner hoods come with installation hardware. Island hoods require mounting kit 33731-85 which includes four 10-ft (3 m) long suspension rods. Other rod lengths are available — contact an Application Specialist for more information.
Note: Canopy hoods require exhausting to the outside or into existing ventilation. Exhaust blower not included.
813,000.00 - ₹951,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Description
Availability Pricing
COMPARE
18 36 Ducted Wall Canopy Fume Hood, 36" W
₹951,000.00INR / Each
COMPARE
18 72 Ducted Wall Canopy Fume Hood, 72" W
Call For Price
COMPARE
18 36 Ducted Island Canopy Fume Hood, 36" W
₹813,000.00INR / Each