Microcaps Bulb Assembly for Disposable Micropipettes
Representative image only.
Microcaps Bulb Assembly for Disposable Micropipettes
Microcaps | Mfr: 1-000-9000| Item: 6115154
Optimize performance of your micropipettes
₹610.00INR / Each
₹610.00INR / Each
Buy It Together
Current Item
Microcaps Bulb Assembly for Disposable Micropipettes
Microcaps Bulb Assembly for Disposable Micropipettes
6115154
₹610.00INR / Each
+
Microcaps Disposable Micropipettes, Glass, 0.25 µL; 100/Pk
0795000
₹6,200.00INR / Pkg of 100
=
Total Price 6810.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Compatible with the MicroCaps® Disposable Micropipettes
More About this Item
Required for dispensing the contents of Drummond MicroCaps®, the Bulb Assembly offers users a simple, yet effective dispensing experience. With a capillary inserted into the amber end of the assembly, MicroCaps® utilizes capillary action to aspirate samples. To dispense, squeeze the bulb, taking care not to cover the hole at the top of the bulb, as a lack of air flow will result in the sample being aspirated into the assembly. Order micropipettes separately.
Specifications
  • Compatible WithMicroCaps® Disposable Micropipettes
  • DescriptionBulb Assembly for Disposable Micropipettes