Mettler Toledo Reach Pt1000-425 pH Electrode; MultiPin™
Representative image only.
Mettler Toledo Reach Pt1000-425 pH Electrode; MultiPin™
Mettler Toledo | Mfr: 51343062| Item: 3590075
Expertly-engineered probes to cover a wide range of applications
₹253,000.00INR / Each
₹253,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo Reach Pt1000-425 pH Electrode; MultiPin™
Mettler Toledo Reach Pt1000-425 pH Electrode; MultiPin™
3590075
₹253,000.00INR / Each
+
Cole-Parmer Adjustable-Volume Pipettor, 2000 to 10000 µL
2450110
₹8,400.00INR / Each
+ + + + =
Total Price 580000.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Cable not included⌡HA glass membrane for high pH and temperature applications
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application, meter, and ATC connection—BNC, waterproof BNC, DIN, and Lemo OO cables available with banana or RCA (cinch) connectors. High alkali glass membrane provides low-alkali error—ideal for high pH and high temperature applications.
Specifications
  • ApplicationsSamples in deep containers
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeHA - High alkalifor high pH and high temp applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC Element1000Ω Pt RTD
  • ConnectorMultipin™
  • Diameter (mm)12
  • Length (mm)425
  • DescriptionpH Electrode; MultiPin™