Mettler Toledo HX204 Halogen Moisture Analyzer
Representative image only.
Mettler Toledo HX204 Halogen Moisture Analyzer
Mettler Toledo | Mfr: 30019875| Item: 1133788
Fast, easy and reliable way to get results of moisture content
₹5,036,000.00INR / Each
for an additional
₹27,400.00INR / Each
for an additional
₹40,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹5,036,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo HX204 Halogen Moisture Analyzer
Mettler Toledo HX204 Halogen Moisture Analyzer
1133788
₹5,036,000.00INR / Each
+ + =
Total Price 5128400.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Fast halogen heating combined with exceptional weighing performance produce highly reliable and repeatable results
  • Preheating, multiple drying programs and more enable accurate measurement of moisture content
  • Superior weighing performance with high 0.001% MC readability
  • Hanging weighing pan eliminates adverse thermal effects on the weighing cell
  • Easy to navigate user interface with large color touchscreen
  • Built-in tests ensure the machine is always performing at its best
  • 3 month warranty
More About this Item
Designed to meet the strict demands of research, quality control and production, this durable Halogen Moisture Analyzer offers high accuracy and repeatability of content moisture results, quickly. It features powerful graphics and what’s dubbed the One Click™ user interface that makes the machine easy to use. Another highlight is real-time drying curves and control charts, in addition to its full compliance, making it an ideal analyzer for use in highly regulated industries.
For easy reporting, connect the analyzer to a computer or printer using a Wi-Fi or Ethernet connection.
To top it off, there are a slew of accessories for this top-of-the-line analyzer, ranging from an anti-theft device, temperature adjustment kit and sample pan handler to transportation cases, reusable steel weighing pans and filters.
3 Month WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Capacity (g)200
  • Readability (mg)0.1
  • Auto ShutoffAutomatic with five switch-off criteria
  • Heater TypeHalogen
  • DescriptionHalogen Moisture Analyzer
  • Warranty3 Months